Learn More
Med Vet International 3 Prongs Retriever – Round, 2.3mm, 2300mm L, J1639A, Each
Catalog No. 5000585692
Encompass_Preferred
Retriever 3-Prong Round
Catalog No. 50-005-85692 Supplier Med Vet International Supplier No. JOR1639A
Retriever 3-Prong Round