Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Propylidenephthalide 96.0+%, TCI America™
SDP

Catalog No. P182425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P182425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P182425G Supplier TCI America Supplier No. P182425G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17369-59-4
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00051813,MFCD00051813
InChI Key NGSZDVVHIGAMOJ-YHYXMXQVSA-N
Synonym 3-propylidenephthalide, 3-propylidene-1 3h-isobenzofuranone, unii-0vb6659n9s, propylidenephthalide, propylidene phthalide, 3e-3-propylidene-2-benzofuran-1-one, 3-propylidene phthalide, phthalide, 3-propylidene 6ci,8ci, 3e-3-propylidene-2-benzofuran-1 3h-one, fema no. 2952
PubChem CID 6259976
IUPAC Name (3Z)-3-propylidene-1,3-dihydro-2-benzofuran-1-one
SMILES CC\C=C1/OC(=O)C2=CC=CC=C12

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 170°C
Quantity 25 g
Formula Weight 174.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Propylidenephthalide
EINECSNumber (5)-0214
RTECSNumber NP7235000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.