Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Propylthiophene 97.0+%, TCI America™
SDP

Catalog No. P11285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11285G 5 g
P11281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11285G Supplier TCI America Supplier No. P11285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1518-75-8
Molecular Formula C7H10S
Molecular Weight (g/mol) 126.22
MDL Number MFCD00143183
InChI Key QZNFRMXKQCIPQY-UHFFFAOYSA-N
PubChem CID 519073
IUPAC Name 3-propylthiophene
SMILES CCCC1=CSC=C1

Specifications

Color Yellow
Boiling Point 161°C
Quantity 5 g
UN Number 1993
Formula Weight 126.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Propylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.