Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Propylxanthine | 41078-02-8 | MFCD00043205 | 98.7% | 194.19 | C8H10N4O2 | 5 MG
SDP

Catalog No. 5000223207
Encompass_Preferred

Enprofylline is a xanthine-derived research compound that functions as a selective and competitive adenosine A2B receptor antagonist and a phosphodiesterase inhibitor. It is provided as a solid for laboratory use and has defined solubility, purity, and storage properties appropriate for in vitro and in vivo research applications.

  • Acts as an adenosine A2B receptor antagonist and phosphodiesterase inhibitor.
  • Molecular formula C8H10N4O2; molecular weight 194.19.
  • High purity (98.7%) suitable for research use.
  • White to light yellow solid; pack size 5 MG.
  • Soluble in DMSO at 62.5 MG/ML; ultrasonic assistance may be required.
  • Store solid at 4°C protected from light; in solution store -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-23207 Supplier Medchemexpress LLC Supplier No. HY141175MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enprofylline is a xanthine-derived research compound that functions as a selective and competitive adenosine A2B receptor antagonist and a phosphodiesterase inhibitor. It is provided as a solid for laboratory use and has defined solubility, purity, and storage properties appropriate for in vitro and in vivo research applications.

  • Acts as an adenosine A2B receptor antagonist and phosphodiesterase inhibitor.
  • Molecular formula C8H10N4O2; molecular weight 194.19.
  • High purity (98.7%) suitable for research use.
  • White to light yellow solid; pack size 5 MG.
  • Soluble in DMSO at 62.5 MG/ML; ultrasonic assistance may be required.
  • Store solid at 4°C protected from light; in solution store -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.