Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyrid-3-ylbenzoic acid, 95%, Thermo Scientific™

Catalog No. CC39801CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC39801CB 250 mg
CC39801DA 1 g
CC39801DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC39801CB Supplier Thermo Scientific Chemicals Supplier No. CC39801CB
Add to Cart

Chemical Identifiers

CAS 4385-77-7
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD03426512
InChI Key PXASTGBSGPFLFJ-UHFFFAOYSA-N
Synonym 3-pyridin-3-yl-benzoic acid, 3-pyridin-3-yl benzoic acid, 3-3'-pyridyl benzoic acid, 3-3-pyridinyl benzoic acid, 3-pyrid-3-ylbenzoic acid, 3-3-pyridyl benzoic acid, benzoic acid, 3-3-pyridinyl, acmc-1bmxh, 3-3'-pyridyl benzoicacid, 3-3-carboxyphenyl pyridine
PubChem CID 2795575
IUPAC Name 3-pyridin-3-ylbenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=CN=CC=C2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 3-Pyrid-3-ylbenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.