Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pyridin-3-ylpropyl (2S)-1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate | 186452-09-5 | MFCD00953146 | 99.8% | 360.4 g/mol | C20H28N2O4 | 50MG
SDP

Catalog No. 5000239379
Encompass_Preferred

GPI-1046 is a small-molecule immunophilin ligand reported to have neuroprotective effects and to modulate glutamate transporter activity. It is supplied in small laboratory pack sizes, typically as a DMSO solution, for preclinical research use only.

  • Chemical formula C20H28N2O4.
  • Molecular weight 360.4 g/mol.
  • CAS number 186452-09-5.
  • Purity 99.8%.
  • Available as solution in DMSO and multiple small pack sizes.
  • Intended for preclinical research use only.

Catalog No. 50-002-39379 Supplier Medchemexpress LLC Supplier No. HY12461950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GPI-1046 is a small-molecule immunophilin ligand reported to have neuroprotective effects and to modulate glutamate transporter activity. It is supplied in small laboratory pack sizes, typically as a DMSO solution, for preclinical research use only.

  • Chemical formula C20H28N2O4.
  • Molecular weight 360.4 g/mol.
  • CAS number 186452-09-5.
  • Purity 99.8%.
  • Available as solution in DMSO and multiple small pack sizes.
  • Intended for preclinical research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.