Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pyridin-3-ylpropyl (2S)-1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate | 186452-09-5 | MFCD00953146 | 99.8% | 360.45 g/mol | C20H28N2O4 | 10MG
SDP

Catalog No. 5000237855
Encompass_Preferred

GPI-1046 is a non-immunosuppressive immunophilin ligand used in research for its neuroprotective effects. It has been reported to upregulate glutamate transporter 1 (GLT1), reduce ethanol intake, and attenuate HIV-associated sensory neuropathy by modulating store-operated calcium entry. The compound is provided as a high-purity liquid (formula C20H28N2O4, MW ~360.45 g/mol).

  • Non-immunosuppressive immunophilin ligand
  • Reported neuroprotective activity
  • Upregulates glutamate transporter 1 (GLT1)
  • Attenuates HIV-associated sensory neuropathy via store-operated calcium entry modulation
  • High purity (≈99.8%)
  • Liquid appearance: colorless to light yellow
  • Chemical formula C20H28N2O4; molecular weight ~360.45 g/mol
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month)

Catalog No. 50-002-37855 Supplier Medchemexpress LLC Supplier No. HY12461910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GPI-1046 is a non-immunosuppressive immunophilin ligand used in research for its neuroprotective effects. It has been reported to upregulate glutamate transporter 1 (GLT1), reduce ethanol intake, and attenuate HIV-associated sensory neuropathy by modulating store-operated calcium entry. The compound is provided as a high-purity liquid (formula C20H28N2O4, MW ~360.45 g/mol).

  • Non-immunosuppressive immunophilin ligand
  • Reported neuroprotective activity
  • Upregulates glutamate transporter 1 (GLT1)
  • Attenuates HIV-associated sensory neuropathy via store-operated calcium entry modulation
  • High purity (≈99.8%)
  • Liquid appearance: colorless to light yellow
  • Chemical formula C20H28N2O4; molecular weight ~360.45 g/mol
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.