Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pyridin-3-ylpropyl (2S)-1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate | 186452-09-5 | MFCD00953146 | 99.8% | 360.45 g/mol | C20H28N2O4 | 10MM 1ML
SDP

Catalog No. 5000235517
Encompass_Preferred

GPI-1046 is a non-immunosuppressive immunophilin ligand used in preclinical research for neuroprotection and modulation of glutamate transport. It is supplied in solution and solid formats for flexible use in in vitro and in vivo studies, and is intended for research use only.

  • Non-immunosuppressive immunophilin ligand with reported neuroprotective activity.
  • Modulates glutamate transporter 1 (GLT1) in preclinical models.
  • Available as a 10 mM solution in DMSO and as solid mg quantities for experimental flexibility.
  • High purity (≈99.8%).
  • Molecular weight 360.45 g/mol; formula C20H28N2O4 for reference.
  • Recommended storage: protect from light; in solvent store at -80°C for long-term preservation.

Catalog No. 50-002-35517 Supplier Medchemexpress LLC Supplier No. HY12461910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GPI-1046 is a non-immunosuppressive immunophilin ligand used in preclinical research for neuroprotection and modulation of glutamate transport. It is supplied in solution and solid formats for flexible use in in vitro and in vivo studies, and is intended for research use only.

  • Non-immunosuppressive immunophilin ligand with reported neuroprotective activity.
  • Modulates glutamate transporter 1 (GLT1) in preclinical models.
  • Available as a 10 mM solution in DMSO and as solid mg quantities for experimental flexibility.
  • High purity (≈99.8%).
  • Molecular weight 360.45 g/mol; formula C20H28N2O4 for reference.
  • Recommended storage: protect from light; in solvent store at -80°C for long-term preservation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.