Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pyridinamine, 6-bromo | 13534-97-9 | MFCD01646116 | 99.5% | 173.01 g/mol | C5H5BrN2 | 100 G
SDP

Catalog No. 5000302017
Encompass_Preferred

6-Bromopyridin-3-amine is a synthetic small-molecule intermediate used in medicinal chemistry and life-science research. It serves as a building block and reagent for the preparation of heterocyclic compounds and for small-molecule synthesis.

  • High purity suitable for research (99.5%).
  • Molecular weight 173.01 g/mol.
  • Chemical formula C5H5BrN2.
  • Available in laboratory-scale package sizes, including 100 g.
  • Certificate of analysis provided for each lot.
  • Useful as a building block in medicinal chemistry and organic synthesis.

Catalog No. 50-003-02017 Supplier Medchemexpress LLC Supplier No. HYW016849100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Bromopyridin-3-amine is a synthetic small-molecule intermediate used in medicinal chemistry and life-science research. It serves as a building block and reagent for the preparation of heterocyclic compounds and for small-molecule synthesis.

  • High purity suitable for research (99.5%).
  • Molecular weight 173.01 g/mol.
  • Chemical formula C5H5BrN2.
  • Available in laboratory-scale package sizes, including 100 g.
  • Certificate of analysis provided for each lot.
  • Useful as a building block in medicinal chemistry and organic synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.