Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Pyridineacetic acid | 501-81-5 | 137.14 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW004515

Encompass_Preferred

3-Pyridineacetic acid is an endogenous metabolite and a higher homologue of nicotinic acid. It is identified as a breakdown product of nicotine and other tobacco alkaloids.

  • Formula: C7H7NO2
  • Appearance: Solid, typically white to off-white
  • Structure classification: Ketones, aldehydes, acids
  • Potential application: Research on the metabolism of nicotine and tobacco alkaloids.

Catalog No. 50-004-49301


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.