Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pyridineacetic acid | 501-81-5 | MFCD00066302 | 100.0% | 137.14 g/mol | C7H7NO2 | 500 MG
SDP

Catalog No. 5000301166
Encompass_Preferred

3-Pyridineacetic acid (2-(pyridin-3-yl)acetic acid) is a small aromatic carboxylic acid used as an endogenous metabolite standard and a reagent in biochemical and pharmacological research. It is supplied at high purity and is suitable for analytical, reference, and preparative applications.

  • High purity suitable for analytical use.
  • Useful as an endogenous metabolite standard in metabolic studies.
  • Aromatic carboxylic acid with pyridine ring chemistry.
  • Molecular weight 137.14 g/mol.
  • Molecular formula C7H7NO2.

Catalog No. 50-003-01166 Supplier Medchemexpress LLC Supplier No. HYW015806500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Pyridineacetic acid (2-(pyridin-3-yl)acetic acid) is a small aromatic carboxylic acid used as an endogenous metabolite standard and a reagent in biochemical and pharmacological research. It is supplied at high purity and is suitable for analytical, reference, and preparative applications.

  • High purity suitable for analytical use.
  • Useful as an endogenous metabolite standard in metabolic studies.
  • Aromatic carboxylic acid with pyridine ring chemistry.
  • Molecular weight 137.14 g/mol.
  • Molecular formula C7H7NO2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.