Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Pyridinecarboxamide, 6-fluoro-N-[2-fluoro-3-[[[2-iodo-4-[1,2,2,2-tetrafluoro-1-(trifluorometh | 1331922-53-2 | 98.3% | 715.23 | 50 MG
SDP

Catalog No. 5000350809
Encompass_Preferred

Modoflaner is an isophenylamide insecticide intended for research use only. It may act through allosteric regulation of gamma-aminobutyric acid-gated chloride channels.

  • Isophenylamide insecticide
  • May act through allosteric regulation of gamma-aminobutyric acid-gated chloride channels
  • Effectively kills moth, xylophone, and gray planthopper
  • Prevents female adult ticks from laying eggs or eggs from hatching
  • White to off-white solid appearance
  • Purity of 98.26%
  • Store at 4°C, protected from light; in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-003-50809 Supplier Medchemexpress LLC Supplier No. HY13744550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Modoflaner is an isophenylamide insecticide intended for research use only. It may act through allosteric regulation of gamma-aminobutyric acid-gated chloride channels.

  • Isophenylamide insecticide
  • May act through allosteric regulation of gamma-aminobutyric acid-gated chloride channels
  • Effectively kills moth, xylophone, and gray planthopper
  • Prevents female adult ticks from laying eggs or eggs from hatching
  • White to off-white solid appearance
  • Purity of 98.26%
  • Store at 4°C, protected from light; in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.