Learn More
Medchemexpress LLC 3-Pyridinecarboxylic acid, 1,2,5,6-tetrahydro-1-methyl-, methyl ester, hydrobromide | 300-08-3 | 99.95% | 1 G
SDP
Catalog No. 5000405530
Encompass_Preferred
Arecoline hydrobromide is a naturally occurring psychoactive alkaloid that is brain-penetrant and orally active. It acts as a partial agonist of nicotinic and muscarinic acetylcholine receptors. It exhibits stimulation, alertness, anxiolysis, and anti-parasitic effects, and can also induce oxidative stress. It is intended for research use only.
  • Acts as a partial agonist of nicotinic and muscarinic acetylcholine receptors.
  • Exhibits stimulation, alertness, anxiolysis, and anti-parasitic effects.
  • Can induce oxidative stress.
  • Induces reactive oxygen species generation and G1/G0 phase cell cycle arrest in HaCaT cells.
  • Alleviates anxiety and depression in CFA-induced mice.
Catalog No. 50-004-05530 Supplier Medchemexpress LLC Supplier No. HYB04891G
Arecoline hydrobromide is a naturally occurring psychoactive alkaloid that is brain-penetrant and orally active. It acts as a partial agonist of nicotinic and muscarinic acetylcholine receptors. It exhibits stimulation, alertness, anxiolysis, and anti-parasitic effects, and can also induce oxidative stress. It is intended for research use only.
  • Acts as a partial agonist of nicotinic and muscarinic acetylcholine receptors.
  • Exhibits stimulation, alertness, anxiolysis, and anti-parasitic effects.
  • Can induce oxidative stress.
  • Induces reactive oxygen species generation and G1/G0 phase cell cycle arrest in HaCaT cells.
  • Alleviates anxiety and depression in CFA-induced mice.