Learn More
Medchemexpress LLC 3-pyridinecarboxylic acid, 1,2,5,6-tetrahydro-, hydrochloride | 6027-91-4 | MFCD00055191 | 99.9% | 163.60 g/mol | C6H10ClNO2 | 50 MG
SDP
Catalog No. 5000212286
Encompass_Preferred
Guvacine hydrochloride is the hydrochloride salt of guvacine, a pyridine alkaloid that inhibits GABA transporters. Supplied for research use to study GABA uptake and transporter pharmacology, it has reported activity across cloned human and rat GAT subtypes.
  • Inhibits GABA transporter activity; reported IC50s include 14 μM (human GAT-1).
  • High purity (≈99.9%) confirmed by COA.
  • Molecular weight 163.60 g/mol; molecular formula C6H10ClNO2.
  • Available in multiple pack sizes, including 50 mg, and solution formats in DMSO.
  • Suitable for biochemical assays and transporter studies.
  • Supplied with analytical documentation such as datasheet and COA.
Catalog No. 50-002-12286 Supplier Medchemexpress LLC Supplier No. HY10080950MG
Guvacine hydrochloride is the hydrochloride salt of guvacine, a pyridine alkaloid that inhibits GABA transporters. Supplied for research use to study GABA uptake and transporter pharmacology, it has reported activity across cloned human and rat GAT subtypes.
  • Inhibits GABA transporter activity; reported IC50s include 14 μM (human GAT-1).
  • High purity (≈99.9%) confirmed by COA.
  • Molecular weight 163.60 g/mol; molecular formula C6H10ClNO2.
  • Available in multiple pack sizes, including 50 mg, and solution formats in DMSO.
  • Suitable for biochemical assays and transporter studies.
  • Supplied with analytical documentation such as datasheet and COA.