Learn More
Medchemexpress LLC 3-Pyridinecarboxylic acid, 2-methyl-, ethyl ester | 1721-26-2 | 99.4% | 165.19 | 5 G
SDP
Catalog No. 5000453582
Encompass_Preferred
Ethyl 2-methylnicotinate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It has been utilized in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, and ethyl 2-(diethylaminomethyl)benzoate.
  • Biochemical reagent for life science research.
  • Can be used as a biological material or organic compound.
  • Suitable for the preparation of amino esters.
Catalog No. 50-004-53582 Supplier Medchemexpress LLC Supplier No. HYW0101055G
Ethyl 2-methylnicotinate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It has been utilized in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, and ethyl 2-(diethylaminomethyl)benzoate.
  • Biochemical reagent for life science research.
  • Can be used as a biological material or organic compound.
  • Suitable for the preparation of amino esters.