Learn More
Medchemexpress LLC 3-pyridinecarboxylic acid, 6-[5-(ethoxycarbonyl)-2-thiazolyl]-, ethyl ester | 1821370-70-0 | MFCD31715412 | 99.9% | 306.34 | C14H14N2O4S | 100 MG
SDP
Catalog No. 5000246144
Encompass_Preferred
Diethyl-pythiDC is a research-use small-molecule inhibitor of collagen prolyl 4-hydroxylases (CP4Hs). It inhibits CP4H activity in cultured cells at concentrations that do not induce iron-deficiency markers and shows low cytotoxicity in tested cell lines, making it useful for in vitro studies of collagen biosynthesis and related pathways.
  • Inhibits collagen prolyl 4-hydroxylase activity in cultured cells.
  • Does not induce iron-deficiency markers under tested conditions.
  • Low cytotoxicity in tested cell lines, including MDA-MB-231.
  • High purity: 99.9% (HPLC).
  • Available as solid (for example, 100 MG) and as a 10 mM, 1 ML solution in DMSO.
Catalog No. 50-002-46144 Supplier Medchemexpress LLC Supplier No. HY103068100MG
Diethyl-pythiDC is a research-use small-molecule inhibitor of collagen prolyl 4-hydroxylases (CP4Hs). It inhibits CP4H activity in cultured cells at concentrations that do not induce iron-deficiency markers and shows low cytotoxicity in tested cell lines, making it useful for in vitro studies of collagen biosynthesis and related pathways.
  • Inhibits collagen prolyl 4-hydroxylase activity in cultured cells.
  • Does not induce iron-deficiency markers under tested conditions.
  • Low cytotoxicity in tested cell lines, including MDA-MB-231.
  • High purity: 99.9% (HPLC).
  • Available as solid (for example, 100 MG) and as a 10 mM, 1 ML solution in DMSO.