Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pyridinecarboxylic acid, chromium(3+) salt (3:1) | 64452-96-6 | MFCD00186328 | ≥98.0% | 418.30 | C18H12CrN3O6 | 1 KG
SDP

Catalog No. 5000284627
Encompass_Preferred

Chromium nicotinate is the nicotinate salt of trivalent chromium used as a dietary supplement and research compound. It is investigated for support of glucose metabolism and type 2 diabetes and is supplied as a solid with molecular formula C18H12CrN3O6 and molecular weight 418.30 g/mol.

  • Used to support glucose metabolism and insulin sensitivity.
  • High purity ≥98.0% (assay).
  • Chemical formula C18H12CrN3O6 and molecular weight 418.30 g/mol.
  • CAS number 64452-96-6 for unambiguous identification.
  • Available in kilogram quantities for bulk laboratory or formulation use.

Catalog No. 50-002-84627 Supplier Medchemexpress LLC Supplier No. HYW5871011KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromium nicotinate is the nicotinate salt of trivalent chromium used as a dietary supplement and research compound. It is investigated for support of glucose metabolism and type 2 diabetes and is supplied as a solid with molecular formula C18H12CrN3O6 and molecular weight 418.30 g/mol.

  • Used to support glucose metabolism and insulin sensitivity.
  • High purity ≥98.0% (assay).
  • Chemical formula C18H12CrN3O6 and molecular weight 418.30 g/mol.
  • CAS number 64452-96-6 for unambiguous identification.
  • Available in kilogram quantities for bulk laboratory or formulation use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.