Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pyridinecarboxylic acid, chromium(3+) salt (3:1) | 64452-96-6 | MFCD00186328 | ≥98.0% | 418.30 | C18H12CrN3O6 | 25 G
SDP

Catalog No. 5000283907
Encompass_Preferred

Chromium nicotinate is the trivalent chromium salt of nicotinic acid supplied for research applications and investigated as a nutritional supplement in type 2 diabetes studies. It is offered as a solid laboratory reagent for analytical and experimental use; consult the Certificate of Analysis for batch-specific handling and storage instructions.

  • Trivalent chromium nicotinate salt.
  • CAS number 64452-96-6.
  • Molecular formula C18H12CrN3O6.
  • Molecular weight 418.30.
  • Appearance off-white to gray solid.
  • Purity ≥98.0%.
  • Storage: room temperature under nitrogen; in solvent -80°C for 2 years, -20°C for 1 year.
  • Intended for research use only; not for human consumption.
  • Investigated as a nutritional supplement for type 2 diabetes research.

Catalog No. 50-002-83907 Supplier Medchemexpress LLC Supplier No. HYW58710125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromium nicotinate is the trivalent chromium salt of nicotinic acid supplied for research applications and investigated as a nutritional supplement in type 2 diabetes studies. It is offered as a solid laboratory reagent for analytical and experimental use; consult the Certificate of Analysis for batch-specific handling and storage instructions.

  • Trivalent chromium nicotinate salt.
  • CAS number 64452-96-6.
  • Molecular formula C18H12CrN3O6.
  • Molecular weight 418.30.
  • Appearance off-white to gray solid.
  • Purity ≥98.0%.
  • Storage: room temperature under nitrogen; in solvent -80°C for 2 years, -20°C for 1 year.
  • Intended for research use only; not for human consumption.
  • Investigated as a nutritional supplement for type 2 diabetes research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.