Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pyridinecarboxylic acid, chromium(3+) salt (3:1) | 64452-96-6 | MFCD00186328 | ≥98.0% | 418.30 g/mol | C18H12CrN3O6 | 50 G
SDP

Catalog No. 5000285134
Encompass_Preferred

Chromium nicotinate is the nicotinate salt of trivalent chromium (Cr3+) supplied as a research-grade powder. It is commonly used in studies of glucose metabolism and as a model nutritional supplement for type 2 diabetes research. The compound's chemical identifiers include CAS 64452-96-6, formula C18H12CrN3O6, and molecular weight 418.30 g/mol.

  • High purity: ≥98.0%
  • Powder form suitable for formulation and analytical studies
  • Used in glucose metabolism and diabetic research
  • Chemical formula C18H12CrN3O6
  • Molecular weight 418.30 g/mol
  • Available in laboratory-scale pack sizes for research use

Catalog No. 50-002-85134 Supplier Medchemexpress LLC Supplier No. HYW58710150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromium nicotinate is the nicotinate salt of trivalent chromium (Cr3+) supplied as a research-grade powder. It is commonly used in studies of glucose metabolism and as a model nutritional supplement for type 2 diabetes research. The compound's chemical identifiers include CAS 64452-96-6, formula C18H12CrN3O6, and molecular weight 418.30 g/mol.

  • High purity: ≥98.0%
  • Powder form suitable for formulation and analytical studies
  • Used in glucose metabolism and diabetic research
  • Chemical formula C18H12CrN3O6
  • Molecular weight 418.30 g/mol
  • Available in laboratory-scale pack sizes for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.