Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Pyridinemethanol | 100-55-0 | 99.58% | 25 G
SDP

Catalog No. 5000406582
Encompass_Preferred

3-Pyridinemethanol, also known as Nicotinyl alcohol, is a pyridine derivative and a cholesterol-lowering agent. It appears as a light yellow to light brown liquid with a molecular weight of 109.13. This compound can be selectively oxidized to vitamin B3 and is intended for laboratory use and the manufacture of other substances.

  • Pyridine derivative
  • Cholesterol-lowering agent
  • Selectively oxidized to vitamin B3
  • Molecular weight of 109.13
  • Chemical formula C6H7NO
  • Appears as a liquid, light yellow to light brown
  • Purity of 99.58%
  • Pure form stable for 3 years at -20°C or 2 years at 4°C
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C

Catalog No. 50-004-06582 Supplier Medchemexpress LLC Supplier No. 25GHYB0893
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Pyridinemethanol, also known as Nicotinyl alcohol, is a pyridine derivative and a cholesterol-lowering agent. It appears as a light yellow to light brown liquid with a molecular weight of 109.13. This compound can be selectively oxidized to vitamin B3 and is intended for laboratory use and the manufacture of other substances.

  • Pyridine derivative
  • Cholesterol-lowering agent
  • Selectively oxidized to vitamin B3
  • Molecular weight of 109.13
  • Chemical formula C6H7NO
  • Appears as a liquid, light yellow to light brown
  • Purity of 99.58%
  • Pure form stable for 3 years at -20°C or 2 years at 4°C
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.