Learn More
Medchemexpress LLC 3-Pyridinemethanol | 100-55-0 | 99.6% | 50 G
SDP
Catalog No. 5000408633
Encompass_Preferred
3-Pyridinemethanol (Nicotinyl alcohol), a pyridine derivative, is a cholesterol-lowering agent. It can be selectively oxidized to vitamin B3.
  • Acts as a cholesterol-lowering agent
  • Can be selectively oxidized to vitamin B3
Catalog No. 50-004-08633 Supplier Medchemexpress LLC Supplier No. HYB089350G
3-Pyridinemethanol (Nicotinyl alcohol), a pyridine derivative, is a cholesterol-lowering agent. It can be selectively oxidized to vitamin B3.
  • Acts as a cholesterol-lowering agent
  • Can be selectively oxidized to vitamin B3