Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Pyridylacetic acid hydrochloride

Catalog No. P658005G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P658005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P658005G Supplier Sigma Aldrich Supplier No. P658005G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Inhibiting effect of 3-pentyn-1-ol on the corrosion of iron in 1m hcl has been investigated
  • Reaction of 3-pentyn-1-ol with fe3(co)12 in ch3oh/koh solution yields closed trinuclear hydridic complex (++-h)fe3(co)9(++3-++3-[h3ccc(ch2)(ch2)co]).3-pentyn-1-ol was used in the synthesis of 4,4-bis(1-methyl-1h-indol-3-yl)pentan-1-ol

Specifications

CAS 6419-36-9
Melting Point 161°C to 163°C (lit.)
Molecular Formula C7H7NO2 -+ HCl
Linear Formula C7H7NO2 -+ HCl
MDL Number MFCD00012819
Quantity 5 g
Synonym 3-Pyridineacetic acid hydrochloride
Molecular Weight (g/mol) 173.6
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.