Learn More
Medchemexpress LLC 3-Pyridylacetic acid hydrochloride | 6419-36-9 | 173.60 | 10 G
SDP
Catalog No. 5000448876
Encompass_Preferred
3-Pyridineacetic acid hydrochloride is a high homolog of nicotinic acid and a decomposition product of nicotine (and other tobacco alkaloids). It can be used to synthesize anti-inflammatory agents or as a precursor for herbicides.
  • High homolog of nicotinic acid
  • Decomposition product of nicotine
  • Precursor for herbicides
  • Can be used to synthesize anti-inflammatory agents
  • White to off-white solid appearance
Catalog No. 50-004-48876 Supplier Medchemexpress LLC Supplier No. HYW00451510G
3-Pyridineacetic acid hydrochloride is a high homolog of nicotinic acid and a decomposition product of nicotine (and other tobacco alkaloids). It can be used to synthesize anti-inflammatory agents or as a precursor for herbicides.
  • High homolog of nicotinic acid
  • Decomposition product of nicotine
  • Precursor for herbicides
  • Can be used to synthesize anti-inflammatory agents
  • White to off-white solid appearance