Learn More
Medchemexpress LLC 3-pyridylamide oxime | 1594-58-7 | >98.0% | 137.14 g·mol⁻¹ | C6H7N3O | 50 G
SDP
Catalog No. 5000300301
Encompass_Preferred
3-Pyridylamide oxime is a pyridine-derived amidoxime used as a synthetic intermediate in pharmaceutical and organic chemistry. It is provided as a solid research reagent and is used for building heterocycles, introducing amidoxime functionality, and advancing medicinal chemistry workflows.
  • Chemical identity: pyridine-based amidoxime (C6H7N3O), CAS 1594-58-7.
  • Molecular weight 137.14 g·mol⁻¹.
  • High purity suitable for synthesis (>98% typical).
  • Useful for medicinal chemistry and pharmaceutical intermediate synthesis.
  • Supplied as a solid, compatible with common organic solvents.
  • Available in multiple pack sizes to support small-scale and scale-up work.
Catalog No. 50-003-00301 Supplier Medchemexpress LLC Supplier No. HYW01580550G
3-Pyridylamide oxime is a pyridine-derived amidoxime used as a synthetic intermediate in pharmaceutical and organic chemistry. It is provided as a solid research reagent and is used for building heterocycles, introducing amidoxime functionality, and advancing medicinal chemistry workflows.
  • Chemical identity: pyridine-based amidoxime (C6H7N3O), CAS 1594-58-7.
  • Molecular weight 137.14 g·mol⁻¹.
  • High purity suitable for synthesis (>98% typical).
  • Useful for medicinal chemistry and pharmaceutical intermediate synthesis.
  • Supplied as a solid, compatible with common organic solvents.
  • Available in multiple pack sizes to support small-scale and scale-up work.