Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Pyrrolidinemethanol, (3R)- | 110013-18-8 | 99.96% | C5H11NO | 1 G
SDP

Catalog No. 5000434742
Encompass_Preferred

(3R)-3-Pyrrolidinemethanol is a molecular building block and PROTAC linker intended for testing and research purposes only. It is not validated for medical applications and cannot be used as pharmaceuticals, foods, or household goods.

  • High purity 99.96%
  • Molecular formula C5H11NO
  • Molecular weight 101.15
  • Appearance: Colorless to off-white liquid
  • Store at room temperature, protected from light
  • In solvent, store at -80°C for 1 year or -20°C for 6 months

Catalog No. 50-004-34742 Supplier Medchemexpress LLC Supplier No. HY602631G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(3R)-3-Pyrrolidinemethanol is a molecular building block and PROTAC linker intended for testing and research purposes only. It is not validated for medical applications and cannot be used as pharmaceuticals, foods, or household goods.

  • High purity 99.96%
  • Molecular formula C5H11NO
  • Molecular weight 101.15
  • Appearance: Colorless to off-white liquid
  • Store at room temperature, protected from light
  • In solvent, store at -80°C for 1 year or -20°C for 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.