Learn More
Medchemexpress LLC 3-Pyrrolidinesulfonic acid, 1-(acetyloxy)-2,5-dioxo- | 152305-87-8 | 98.0% | C6H7NO7S | 25 MG
SDP
Catalog No. 5000352611
Encompass_Preferred
Sulfo-NHS-Acetate is an alkyl chain-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs are compounds containing two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Targets an E3 ubiquitin ligase
  • Targets a specific protein
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
  • Relevant to cancer research
  • Relevant to cancer targeted therapy
  • Classified as a PROTAC linker
  • Functions as an inhibitor
Catalog No. 50-003-52611 Supplier Medchemexpress LLC Supplier No. 25MGHY140341
Sulfo-NHS-Acetate is an alkyl chain-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs are compounds containing two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Targets an E3 ubiquitin ligase
  • Targets a specific protein
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
  • Relevant to cancer research
  • Relevant to cancer targeted therapy
  • Classified as a PROTAC linker
  • Functions as an inhibitor