Learn More
Medchemexpress LLC 3-pyrrolidinol, 1-(phenylmethyl)-, (3S)- | 101385-90-4 | MFCD00075485 | 99.9% | 177.25 g/mol | C11H15NO | 5 G
SDP
Catalog No. 5000303333
Encompass_Preferred
(S)-1-Benzylpyrrolidin-3-ol is a chiral pyrrolidinyl alcohol used as a synthetic intermediate and analytical reference in medicinal chemistry. It is supplied as the S-enantiomer with high chemical purity and is employed for small-scale synthesis, impurity characterization, and method development related to antihypertensive compound impurities.
  • Chiral S-enantiomer for stereospecific synthesis.
  • High chemical purity: 99.9%.
  • Molecular formula C11H15NO, molecular weight 177.25 g/mol.
  • Available in small research quantities suitable for analytical and synthetic work.
  • Stable under recommended storage conditions for multi-year shelf life.
  • Suitable as an intermediate and impurity standard in pharmaceutical research.
Catalog No. 50-003-03333 Supplier Medchemexpress LLC Supplier No. HYW0166445G
(S)-1-Benzylpyrrolidin-3-ol is a chiral pyrrolidinyl alcohol used as a synthetic intermediate and analytical reference in medicinal chemistry. It is supplied as the S-enantiomer with high chemical purity and is employed for small-scale synthesis, impurity characterization, and method development related to antihypertensive compound impurities.
  • Chiral S-enantiomer for stereospecific synthesis.
  • High chemical purity: 99.9%.
  • Molecular formula C11H15NO, molecular weight 177.25 g/mol.
  • Available in small research quantities suitable for analytical and synthetic work.
  • Stable under recommended storage conditions for multi-year shelf life.
  • Suitable as an intermediate and impurity standard in pharmaceutical research.