Learn More
Accela Chembio Inc 3-pyrrolidinol | 1g | 40499-83-0 | MFCD00005256 | 97+% | D: 1.076 | Shelf Life: 1260 Days | Light Sensitive/moisture Sensitive
Catalog No. 50250046
Encompass_Preferred
3-pyrrolidinol | 1g | 40499-83-0 | MFCD00005256 | 97+% | D: 1.076 | Shelf Life: 1260 Days | Light Sensitive/moisture Sensitive
Catalog No. 50-250-046 Supplier Accela Chembio Inc Supplier No. SY0031431G
3-pyrrolidinol | 1g | 40499-83-0 | MFCD00005256 | 97+% | D: 1.076 | Shelf Life: 1260 Days | Light Sensitive/moisture Sensitive