Learn More
Medchemexpress LLC 3-Quinolinecarboxylic acid, 4-hydroxy-8-nitro- | 35973-25-2 | 98.5% | 1 MG
SDP
Catalog No. 5000341822
Encompass_Preferred
DNA2 inhibitor C5 is a potent, competitive, and specific DNA2 nuclease inhibitor with an IC50 of 20 μM. It inhibits the nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2. It can be used in breast cancer and colorectal cancer research.
  • Potent, competitive, and specific DNA2 nuclease inhibitor
  • Inhibits nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2
  • Useful in breast cancer and colorectal cancer research
  • Shows strong synergistic effect in inhibiting MCF7 cell survival with MK4827
Catalog No. 50-003-41822 Supplier Medchemexpress LLC Supplier No. HY1287291MG
DNA2 inhibitor C5 is a potent, competitive, and specific DNA2 nuclease inhibitor with an IC50 of 20 μM. It inhibits the nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2. It can be used in breast cancer and colorectal cancer research.
  • Potent, competitive, and specific DNA2 nuclease inhibitor
  • Inhibits nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2
  • Useful in breast cancer and colorectal cancer research
  • Shows strong synergistic effect in inhibiting MCF7 cell survival with MK4827