Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-quinolinecarboxylic acid, 7-[(3S,5S)-3-amino-5-methyl-1-piperidinyl]-1-cyclopropyl-1,4-dihydro-8-methoxy-4-oxo | 99.9% | 374.45 | 5 MG
SDP

Catalog No. 5000360021
Encompass_Preferred

Nemonoxacin-d3 is a deuterated analog of Nemonoxacin, suitable for research applications. This product is supplied as an off-white to light yellow solid, confirmed by testing to comply with its specifications.

  • Chemical name: 3-quinolinecarboxylic acid, 7-[(3S,5S)-3-amino-5-methyl-1-piperidinyl]-1-cyclopropyl-1,4-dihydro-8-methoxy-4-oxo-d3
  • Molecular formula: C20H22D3N3O4
  • Molecular weight: 374.45
  • Purity (LCMS): 99.93%
  • Appearance: Off-white to light yellow solid
  • Isotopic enrichment: 99.76%
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
  • For research use only

Catalog No. 50-003-60021 Supplier Medchemexpress LLC Supplier No. HY14956S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nemonoxacin-d3 is a deuterated analog of Nemonoxacin, suitable for research applications. This product is supplied as an off-white to light yellow solid, confirmed by testing to comply with its specifications.

  • Chemical name: 3-quinolinecarboxylic acid, 7-[(3S,5S)-3-amino-5-methyl-1-piperidinyl]-1-cyclopropyl-1,4-dihydro-8-methoxy-4-oxo-d3
  • Molecular formula: C20H22D3N3O4
  • Molecular weight: 374.45
  • Purity (LCMS): 99.93%
  • Appearance: Off-white to light yellow solid
  • Isotopic enrichment: 99.76%
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.