Learn More
Medchemexpress LLC 3-Quinolineethanol, 6-bromo-α-[2-(dimethylamino)ethyl]-2-methoxy-α-1-naphthalenyl-β-phenyl-, (αR,βS)-r | 654653-93-7 | 97.6% | 555.50 g·mol⁻¹ | C32H31BrN2O2 | 10 MG
SDP
Catalog No. 5000297407
Encompass_Preferred
rel-Bedaquiline is a drug impurity related to the anti-tuberculosis agent bedaquiline and is supplied as an analytical reference for research use only. It is intended for chromatographic and analytical method development, impurity profiling, and stability studies. Consult the Certificate of Analysis for batch-specific purity and storage conditions.
  • Intended for research use only; not for human or clinical use.
  • Impurity standard for analytical and chromatographic method development.
  • High purity reference: 97.56% by HPLC (batch dependent).
  • Suitable for impurity profiling and stability studies.
  • Brominated quinoline core; molecular weight 555.50 g·mol⁻¹.
Catalog No. 50-002-97407 Supplier Medchemexpress LLC Supplier No. HYI103110MG
rel-Bedaquiline is a drug impurity related to the anti-tuberculosis agent bedaquiline and is supplied as an analytical reference for research use only. It is intended for chromatographic and analytical method development, impurity profiling, and stability studies. Consult the Certificate of Analysis for batch-specific purity and storage conditions.
  • Intended for research use only; not for human or clinical use.
  • Impurity standard for analytical and chromatographic method development.
  • High purity reference: 97.56% by HPLC (batch dependent).
  • Suitable for impurity profiling and stability studies.
  • Brominated quinoline core; molecular weight 555.50 g·mol⁻¹.