Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-3-Quinuclidinyl Benzilate

Catalog No. C0035MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
11 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0035MG 11 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. C0035MG Supplier Sigma Aldrich Supplier No. C0035MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pharmacological agent used as muscarinic M3 antagonist for the inhaled treatment of chronic obstructive pulmonary disease (COPD)Nonselective muscarinic acetylcholine receptor antagonist. More potent enantiomer of QNB.

Specifications

CAS 38285-78-8
Linear Formula C8H5BrF4S
MDL Number MFCD27996155
Quantity 11 Ea.
Synonym (R)-(-)-QNB; (R)-(-)-Quinuclidinyl-alpha-hydroxydiphenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.