Learn More
Medchemexpress LLC 3-((R)-2-carboxypiperazin-4-yl)-propyl-1-phosphonic acid | 126453-07-4 | 99.0% | 252.20 g/mol | C8H17N2O5P | 1 MG
SDP
Catalog No. 5000212410
Encompass_Preferred
(R)-CPP is a research-grade NMDA receptor antagonist provided as a powdered chemical for laboratory neuropharmacology studies. The R-enantiomer selectively blocks NMDA receptor-mediated responses and is supplied for research use only.
  • Highly potent NMDA receptor antagonist for experimental use.
  • High purity (98.99% assay), suitable for research applications.
  • Molecular weight 252.20 g/mol; formula C8H17N2O5P.
  • Available in small, measured quantities for laboratory experiments.
  • Store sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-12410 Supplier Medchemexpress LLC Supplier No. HY1008141MG
(R)-CPP is a research-grade NMDA receptor antagonist provided as a powdered chemical for laboratory neuropharmacology studies. The R-enantiomer selectively blocks NMDA receptor-mediated responses and is supplied for research use only.
  • Highly potent NMDA receptor antagonist for experimental use.
  • High purity (98.99% assay), suitable for research applications.
  • Molecular weight 252.20 g/mol; formula C8H17N2O5P.
  • Available in small, measured quantities for laboratory experiments.
  • Store sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).