Learn More
Medchemexpress LLC 3-((R)-2-carboxypiperazin-4-yl)-propyl-1-phosphonic acid | 126453-07-4 | 99.0% | 252.20 g·mol⁻¹ | C8H17N2O5P | 25 MG
SDP
Catalog No. 5000211777
Encompass_Preferred
(R)-CPP is the R-enantiomer of CPP, a potent competitive N-methyl-D-aspartate (NMDA) receptor antagonist used as a research reagent in neuropharmacology and electrophysiology. It is provided as a high-purity solid for in vitro and in vivo studies and is not for human or clinical use.
  • Highly potent NMDA receptor antagonist suitable for receptor pharmacology.
  • R-enantiomer with greater activity than the racemic mixture.
  • High reported purity (98.99%).
  • Solid form with defined molecular weight and formula (252.20 g·mol⁻¹; C8H17N2O5P).
  • Commonly used in electrophysiology and glutamatergic signaling studies.
Catalog No. 50-002-11777 Supplier Medchemexpress LLC Supplier No. HY10081425MG
(R)-CPP is the R-enantiomer of CPP, a potent competitive N-methyl-D-aspartate (NMDA) receptor antagonist used as a research reagent in neuropharmacology and electrophysiology. It is provided as a high-purity solid for in vitro and in vivo studies and is not for human or clinical use.
  • Highly potent NMDA receptor antagonist suitable for receptor pharmacology.
  • R-enantiomer with greater activity than the racemic mixture.
  • High reported purity (98.99%).
  • Solid form with defined molecular weight and formula (252.20 g·mol⁻¹; C8H17N2O5P).
  • Commonly used in electrophysiology and glutamatergic signaling studies.