Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-((R)-2-carboxypiperazin-4-yl)-propyl-1-phosphonic acid | 126453-07-4 | MFCD00153786 | 99.0% | 252.20 g·mol⁻1 | C8H17N2O5P | 5 MG
SDP

Catalog No. 5000211814
Encompass_Preferred

(R)-CPP is a selective, potent N-methyl-D-aspartate (NMDA) receptor antagonist provided as a research-grade solid for neuroscience applications. It competitively inhibits NMDA receptor-mediated signaling and is used to study glutamatergic neurotransmission, synaptic plasticity, and receptor pharmacology.

  • Potent, selective NMDA receptor antagonist.
  • High reported purity (≈99.0%).
  • Water soluble (≥25.22 mg/mL).
  • Molecular weight 252.20 g·mol⁻1.
  • Solid form, off-white to yellow.
  • CAS number 126453-07-4 for unambiguous identification.

Catalog No. 50-002-11814 Supplier Medchemexpress LLC Supplier No. HY1008145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-CPP is a selective, potent N-methyl-D-aspartate (NMDA) receptor antagonist provided as a research-grade solid for neuroscience applications. It competitively inhibits NMDA receptor-mediated signaling and is used to study glutamatergic neurotransmission, synaptic plasticity, and receptor pharmacology.

  • Potent, selective NMDA receptor antagonist.
  • High reported purity (≈99.0%).
  • Water soluble (≥25.22 mg/mL).
  • Molecular weight 252.20 g·mol⁻1.
  • Solid form, off-white to yellow.
  • CAS number 126453-07-4 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.