Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex 3 Ring Binder Holder
SDP

Catalog No. 22000380
Change view
Click to view available options
Mounting Type:
Powerful Magnet
Wall, Keyhole Slots
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
22-000-380 Wall, Keyhole Slots
22-000-379 Powerful Magnet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-380 Supplier Poltex Supplier No. BINDR250W
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clean room approved, durable, cleanable binder holder.

Specifications

Color Clear
Material PETG
Mounting Type Wall, Keyhole Slots
For Use With (Application) Binders, documents
Quantity 1
Height (Metric) 247 mm
Length (Metric) 89 mm
Width (Metric) 323 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.