Learn More
Chemglass Life Sciences 3 Ring Hose Connection, "A" = 4.0, "B" = 8.0, "C" = 10, "D" = 18; "E" = 10
SDP
Catalog No. 501531984
Encompass_Preferred
3 Ring Hose Connection, "A" = 4.0, "B" = 8.0, "C" = 10, "D" = 18; "E" = 10. Connections are tooled to form a flare for sealing directly to apparatus using the tubing as a holder. Serrations are tooled on both ends of an 8" length of tubing.
Catalog No. 50-153-1984 Supplier Chemglass Life Sciences Supplier No. CGB30101
3 Ring Hose Connection, "A" = 4.0, "B" = 8.0, "C" = 10, "D" = 18; "E" = 10. Connections are tooled to form a flare for sealing directly to apparatus using the tubing as a holder. Serrations are tooled on both ends of an 8" length of tubing.