Learn More
Chemglass Life Sciences Glassblowers Hose Connection, Micro, Regular, 3 Rings, Dimensions (mm): "A" - 3.5, "B" - 8, "C" - 10, "D" - 18
SDP
Catalog No. CG30202
Encompass_Preferred
Glassblowers Hose Connection, Micro, Regular, 3 Rings, Dimensions (mm): "A" - 3.5, "B" - 8, "C" - 10, "D" - 18. Used to construct small scale apparatus. Available in direct sealing or regular style. Serrations are tooled on both ends of an 8in length of tubing. Hose connections are SOLD IN PACKAGES OF 10 SERRATED CONNECTORS (5 pcs. tooled on both ends).
Catalog No. CG30202 Supplier Chemglass Life Sciences Supplier No. CG30202
Glassblowers Hose Connection, Micro, Regular, 3 Rings, Dimensions (mm): "A" - 3.5, "B" - 8, "C" - 10, "D" - 18. Used to construct small scale apparatus. Available in direct sealing or regular style. Serrations are tooled on both ends of an 8in length of tubing. Hose connections are SOLD IN PACKAGES OF 10 SERRATED CONNECTORS (5 pcs. tooled on both ends).