Learn More
Chemglass Life Sciences Glassblowers Hose Connection, Micro, Direct Sealing, 3 Rings, Dimensions (mm): "A" - 3.5, "B" - 8, "C" - 10, "D" - 18
SDP
Supplier:  Chemglass Life Sciences CG30201
Encompass_Preferred
Glassblowers Hose Connection, Micro, Direct Sealing, 3 Rings, Dimensions (mm): "A" - 3.5, "B" - 8, "C" - 10, "D" - 18. Used to construct small scale apparatus. Available in direct sealing or regular style. Serrations are tooled on both ends of an 8in length of tubing. Hose connections are SOLD IN PACKAGES OF 10 SERRATED CONNECTORS (5 pcs. tooled on both ends).
Catalog No. CG30201