Learn More
SKYLINE CENTER Item #W13 Bulk 3" Round - 1,000ct.
Catalog No. NC1590724
Encompass
3” ROUND 100% COTTON PATCHES Uses: Cleaning - guns, firearms, melt flow index, melt plastometers, capillary rheometers, optics, laboratory equipment. Gun Cleaning - 16, 12 and 10 Gauge
Catalog No. NC1590724 Supplier SKYLINE CENTER Supplier No. W13
3” ROUND 100% COTTON PATCHES Uses: Cleaning - guns, firearms, melt flow index, melt plastometers, capillary rheometers, optics, laboratory equipment. Gun Cleaning - 16, 12 and 10 Gauge