Learn More
Ace Glass, Inc. 3 S/C GAS MANIFOLD 3 S/C GAS MANIFOLD
SDP
Catalog No. 50974802
Encompass_Preferred
For portable gas analysis apparatus. Three straight 2 mm bore glass stopcocks and one three-way stopcock with back port, fabricated on capillary tubing.
Catalog No. 50-974-802 Supplier Ace Glass, Inc. Supplier No. 741610
For portable gas analysis apparatus. Three straight 2 mm bore glass stopcocks and one three-way stopcock with back port, fabricated on capillary tubing.