Learn More
Cayman Chemical 3'-Sialyllactose (sodium salt), Purity: ≥98%, CAS: 128596-80-5, 10 mg
SDP
Catalog No. 502966859
Encompass_Preferred
An abundant oligosaccharide in the milk of many mammals, including cows and humans, particularly postpartum; avidly binds several viral strains, including strains of influenza, HIV-1, reovirus, and polyomavirus, in some cases altering viral infectivity
Catalog No. 50-296-6859 Supplier Cayman Chemical Supplier No. 1661710 MG
An abundant oligosaccharide in the milk of many mammals, including cows and humans, particularly postpartum; avidly binds several viral strains, including strains of influenza, HIV-1, reovirus, and polyomavirus, in some cases altering viral infectivity