Learn More
Medchemexpress LLC 3'-Sialyllactose (sodium) | 128596-80-5 | MFCD00166999 | 99.9% | 655.53 | C23H38NNaO19 | 50 MG
SDP
Catalog No. 5000243009
Encompass_Preferred
3'-Sialyllactose (sodium) is the sodium salt of a milk-derived oligosaccharide supplied for research use. It is reported to act as a prebiotic and to modulate immune responses, with documented anti-inflammatory and anti-arthritic effects. The material is provided as a high-purity solid for biochemical, microbiome, and inflammation-related studies.
  • High purity (99.9%) suitable for research
  • Provided as a stable sodium salt solid for handling and storage
  • Useful for prebiotic and microbiome studies
  • Supports research into immune homeostasis and anti-inflammatory effects
  • Well-characterized chemical identity and molecular weight (655.53)
Catalog No. 50-002-43009 Supplier Medchemexpress LLC Supplier No. HY108065A50MG
3'-Sialyllactose (sodium) is the sodium salt of a milk-derived oligosaccharide supplied for research use. It is reported to act as a prebiotic and to modulate immune responses, with documented anti-inflammatory and anti-arthritic effects. The material is provided as a high-purity solid for biochemical, microbiome, and inflammation-related studies.
  • High purity (99.9%) suitable for research
  • Provided as a stable sodium salt solid for handling and storage
  • Useful for prebiotic and microbiome studies
  • Supports research into immune homeostasis and anti-inflammatory effects
  • Well-characterized chemical identity and molecular weight (655.53)