Learn More
Gilson Company Inc 3"SIEVE BR/SS FULL #200
Catalog No. NC2147399
Encompass_Preferred
3" Sieve, Brass/Stainless, Full Height, No. 2003in (76mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 200 (75µm) openings and full height brass frame. The overall height is 1-3/4in and the stacked height is 1-1/8in.
This item is not returnable. View return policy
Catalog No. NC2147399 Supplier Gilson Company Inc Supplier No. V3CF #200
3" Sieve, Brass/Stainless, Full Height, No. 2003in (76mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 200 (75µm) openings and full height brass frame. The overall height is 1-3/4in and the stacked height is 1-1/8in.