Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex 3 Slot Landscape Documet Holder
SDP

Catalog No. 22000439
Change view
Click to view available options
Mounting Type:
Powerful Magnet
Wall, Keyhole Slots
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
22-000-439 Powerful Magnet
22-000-438 Wall, Keyhole Slots
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-439 Supplier Poltex Supplier No. 3UPFILEM
Add to Cart
Add to Cart

Store documents up and out of the way.

Specifications

Product Type Document Holder
Color Clear
Material Acrylic
Mounting Type Powerful Magnet
For Use With (Application) QC Documents, Paper Files
Quantity 1
Height (Metric) 297 mm
Length (Metric) 56 mm
Thickness 0.25 in.
Width (Metric) 305 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.