Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, 3-Stop, Peroxide-Cured Silicone 2.79 mm ID; 6/PK
Catalog No. NC9904017
Encompass
Designed for optimal operation in your Ismatec pumps. It is resistant to ozone and UV light. It won't crack, swell or harden. Translucent yellow color. Temperature range: -50 to 74C (-58 to 165F).
Catalog No. NC9904017 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0762448
Designed for optimal operation in your Ismatec pumps. It is resistant to ozone and UV light. It won't crack, swell or harden. Translucent yellow color. Temperature range: -50 to 74C (-58 to 165F).