Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, 3-Stop, Peroxide-Cured Silicone 1.02 mm ID; 6/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0762428
Encompass
Designed for optimal operation in your Ismatec pumps. It is resistant to ozone and UV light. It won't crack, swell or harden. Translucent yellow color. Temperature range: -50 to 74C (-58 to 165F).
Catalog No. NC9440676