Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing with Flared Ends, 3-Stop, PVC, 0.25 mm ID; 12/PK
Catalog No. NC1856887
Encompass
Three-Stop Flared PVC Color-coded Autoanalysis Tubing for Ismatec Pumps is USP Class VI tubing that is Phthalate-free. It has a long pumping life and exhibits a wide range of acid and chemical resistance, thus making it an ideal choice for Pharmaceutical industries. Plasticizer is very tightly bonded into the material, preventing any leaching of component chemicals into the medium being transported, maintaining its sterility. The tubing is transparent, which allows visual observation of tubing contents. It also offers very low gas permeability and can withstand high pressure contents.
Catalog No. NC1856887 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9761712
Three-Stop Flared PVC Color-coded Autoanalysis Tubing for Ismatec Pumps is USP Class VI tubing that is Phthalate-free. It has a long pumping life and exhibits a wide range of acid and chemical resistance, thus making it an ideal choice for Pharmaceutical industries. Plasticizer is very tightly bonded into the material, preventing any leaching of component chemicals into the medium being transported, maintaining its sterility. The tubing is transparent, which allows visual observation of tubing contents. It also offers very low gas permeability and can withstand high pressure contents.