Learn More
AVANTOR MASTERFLEX (VWR LLC) 3-STP TYGN E-LAB 2.29MMID 12PK
Catalog No. 502741892
Encompass
Masterflex Ismatec Pump Tubing, 3-Stop, Tygon S3™ E-Lab, 2.29 mm ID; 12/PK, Avantor
Catalog No. 50-274-1892 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9646144
Masterflex Ismatec Pump Tubing, 3-Stop, Tygon S3™ E-Lab, 2.29 mm ID; 12/PK, Avantor