Learn More
AVANTOR MASTERFLEX (VWR LLC) 3-STP TYGN E-LFL 0.51MMID 12PK
Catalog No. 502741961
Encompass
Masterflex Ismatec Pump Tubing, 3-Stop, Tygon S3™ E-LFL, 0.51 mm ID; 12/PK, Avantor
Catalog No. 50-274-1961 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9645018
Masterflex Ismatec Pump Tubing, 3-Stop, Tygon S3™ E-LFL, 0.51 mm ID; 12/PK, Avantor